Learn More
Medchemexpress LLC Darolutamide-d4 | 2484757-20-0 | 98.6% | 402.87 | 500 UG
SDP
Catalog No. 5000390482
Encompass_Preferred
Darolutamide-d4 is a deuterium-labeled version of Darolutamide. It is an orally active, competitive androgen receptor (AR) antagonist that inhibits testosterone-induced AR nuclear translocation and transcriptional activation. This compound selectively affects AR-positive cells by inhibiting AR-dependent signaling pathways, and its active metabolite maintains full antagonistic activity against AR mutants. It can be used for research on prostate cancer, including androgen receptor-dependent prostate cancer.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90482 Supplier Medchemexpress LLC Supplier No. HY16985S500UG
Darolutamide-d4 is a deuterium-labeled version of Darolutamide. It is an orally active, competitive androgen receptor (AR) antagonist that inhibits testosterone-induced AR nuclear translocation and transcriptional activation. This compound selectively affects AR-positive cells by inhibiting AR-dependent signaling pathways, and its active metabolite maintains full antagonistic activity against AR mutants. It can be used for research on prostate cancer, including androgen receptor-dependent prostate cancer.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.