Learn More
Medchemexpress LLC Darolutamide-d4 | 2484757-20-0 | 98.6% | 402.87 g/mol | C19H15D4ClN6O2 | 10 MG
SDP
Catalog No. 5000295139
Encompass_Preferred
Darolutamide-d4 is a deuterium-labeled analogue of the androgen receptor antagonist darolutamide supplied as a solid for research use. It is used as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS and reflects the parent compound's AR antagonist profile.
  • Deuterium-labeled internal standard for analytical assays.
  • Intended for research use only; not for human or clinical use.
  • High purity (≈98.6%).
  • Soluble in DMSO at 100 mg/mL; sonication recommended.
  • Powder storage: -20°C (multi-year stability); in solution: -80°C for long-term storage.
  • White to off-white solid; molecular weight ~402.87 g/mol.
Catalog No. 50-002-95139 Supplier Medchemexpress LLC Supplier No. HY16985S10MG
Darolutamide-d4 is a deuterium-labeled analogue of the androgen receptor antagonist darolutamide supplied as a solid for research use. It is used as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS and reflects the parent compound's AR antagonist profile.
  • Deuterium-labeled internal standard for analytical assays.
  • Intended for research use only; not for human or clinical use.
  • High purity (≈98.6%).
  • Soluble in DMSO at 100 mg/mL; sonication recommended.
  • Powder storage: -20°C (multi-year stability); in solution: -80°C for long-term storage.
  • White to off-white solid; molecular weight ~402.87 g/mol.