Learn More
Medchemexpress LLC Darolutamide-d4 (ODM-201-d4) | 2484757-20-0 | 98.6% | 402.87 | 5 MG
SDP
Catalog No. 5000391126
Encompass_Preferred
Darolutamide-d4 (ODM-201-d4) is a deuterium-labeled version of Darolutamide (HY-16985). Darolutamide (ODM-201) is an orally active, competitive androgen receptor (AR) antagonist, with a Ki of 11 nM for rat wild-type AR (wtAR) and an IC50 of 26 nM for human wild-type AR (hAR)-mediated transcriptional activation. It inhibits testosterone-induced AR nuclear translocation and transcriptional activation and is used in research for prostate cancer.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Selectively affects AR-positive cells
  • Maintains full antagonistic activity against AR mutants
Catalog No. 50-003-91126 Supplier Medchemexpress LLC Supplier No. HY16985S5MG
Darolutamide-d4 (ODM-201-d4) is a deuterium-labeled version of Darolutamide (HY-16985). Darolutamide (ODM-201) is an orally active, competitive androgen receptor (AR) antagonist, with a Ki of 11 nM for rat wild-type AR (wtAR) and an IC50 of 26 nM for human wild-type AR (hAR)-mediated transcriptional activation. It inhibits testosterone-induced AR nuclear translocation and transcriptional activation and is used in research for prostate cancer.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Selectively affects AR-positive cells
  • Maintains full antagonistic activity against AR mutants