Learn More
Selleck Chemical LLC Darolutamide ODM-201-5mg
SDP
Catalog No. 502389656
Encompass_Preferred
Darolutamide (ODM-201, BAY-1841788) is a novel androgen receptor (AR) antagonist that blocks AR nuclear translocation with Ki of 11 nM. Phase 3.
Catalog No. 50-238-9656 Supplier Selleck Chemical LLC Supplier No. S75595MG
Darolutamide (ODM-201, BAY-1841788) is a novel androgen receptor (AR) antagonist that blocks AR nuclear translocation with Ki of 11 nM. Phase 3.