Learn More
Medchemexpress LLC HY-101569 2mg , Darovasertib CAS:1874276-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015692MG
Encompass_Preferred
HY-101569 2mg Darovasertib CAS: 1874276-76-2 Darovasertib (LXS196) is a potent, selective and orally active protein kinase C (PKC) inhibitor, with IC50 values of 1.9 nM, 0.4 nM and 3.1 μM for PKCα, PKCθ and GSK3β, respectively. Darovasertib has the potential for uveal melanoma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0407