Learn More
Medchemexpress LLC Darovasertib (LXS196) | 1874276-76-2 | MFCD31544328 | >98% | 472.47 | C22H23F3N8O
SDP
Catalog No. 5000248980
Encompass_Preferred
Darovasertib (LXS196) is a potent, selective, orally active pan-protein kinase C (PKC) inhibitor developed for oncology research, with demonstrated low-nanomolar activity against multiple PKC isoforms. It is provided with characterization data and solubility guidance for preparation of experimental stocks, and is intended for research use in studies such as uveal melanoma models.
  • Potent inhibition of PKC isoforms at low nanomolar concentrations.
  • Selective, orally active small molecule suitable for in vitro and in vivo studies.
  • Characterized molecular formula and molecular weight for analytical confirmation.
  • Supplied with solubility and handling guidance for DMSO stock preparation.
  • High chemical purity appropriate for research applications.
Catalog No. 50-002-48980 Supplier Medchemexpress LLC Supplier No. HY10156925MG
Darovasertib (LXS196) is a potent, selective, orally active pan-protein kinase C (PKC) inhibitor developed for oncology research, with demonstrated low-nanomolar activity against multiple PKC isoforms. It is provided with characterization data and solubility guidance for preparation of experimental stocks, and is intended for research use in studies such as uveal melanoma models.
  • Potent inhibition of PKC isoforms at low nanomolar concentrations.
  • Selective, orally active small molecule suitable for in vitro and in vivo studies.
  • Characterized molecular formula and molecular weight for analytical confirmation.
  • Supplied with solubility and handling guidance for DMSO stock preparation.
  • High chemical purity appropriate for research applications.