Learn More
Cayman Chemical DARPP32 PhosphoThr34 Polyclo
SDP
Catalog No. 502886942
Encompass_Preferred
Immunogen: Phosphopeptide corresponding to amino acid residues surrounding the phospho-Thr34 site of rat DARPP-32 • Host: Rabbit • Species Reactivity: (+) Mouse, Rat • Cross Reactivity: (+) DARPP-32 (phospho-Thr34); (-) Unphosphorylated DARPP-32 • Applications: WB • MW = ~32 kDa
Catalog No. 50-288-6942 Supplier Cayman Chemical Supplier No. 106031 EA
Immunogen: Phosphopeptide corresponding to amino acid residues surrounding the phospho-Thr34 site of rat DARPP-32 • Host: Rabbit • Species Reactivity: (+) Mouse, Rat • Cross Reactivity: (+) DARPP-32 (phospho-Thr34); (-) Unphosphorylated DARPP-32 • Applications: WB • MW = ~32 kDa