Learn More
Enterprise Technology Solutions Dart Container Corp Lids,f/Hot/ColdFoamCups,Straw/Slotted,Lrg,100/PK,10/CT, TLT
SDP
Catalog No. 5000790741
Encompass_Preferred
Translucent lids are perfect for use with foam cups to contain your hot or cold while you're on-the-go. Center features a slot for a straw for added convenience. Lid is designed for use with Dart 32AJ32, 32TJ32, 44AJ32, 44TJ32 and 60AJ32 cups (sold separately). Made of high-impact polystyrene, these quality lids fit tightly on cups to prevent drips or spills. Temperature tolerances ranges from 14 degrees to 203 degrees Fahrenheit.
Catalog No. 50-007-90741 Supplier Enterprise Technology Solutions Supplier No. DCC32SL1
Translucent lids are perfect for use with foam cups to contain your hot or cold while you're on-the-go. Center features a slot for a straw for added convenience. Lid is designed for use with Dart 32AJ32, 32TJ32, 44AJ32, 44TJ32 and 60AJ32 cups (sold separately). Made of high-impact polystyrene, these quality lids fit tightly on cups to prevent drips or spills. Temperature tolerances ranges from 14 degrees to 203 degrees Fahrenheit.