Learn More
AdipoGen Darunavir (10 mg)
SDP
Supplier:  AdipoGen AGCR13712M010
Encompass_Preferred
Chemical. CAS: 206361-99-1. Formula: C27H37N3O7S. MW: 547.7. Darunavir is a highly potent HIV protease inhibitor (IC50=3-6nM, depending on laboratory HIV-1 strain), an antiretroviral medication used to treat and prevent HIV/AIDS. Darunavir is a second generation HIV-1 protease inhibitor that inhibits replication of various laboratory strains and clinical isolates of HIV-1, including those resistant to first generation protease inhibitors. It inhibits cell-free diffusion and cell-to-cell spread of HIV-1 in Jurkat cell populations. Formulations containing darunavir have been used in combination therapy for the treatment of HIV. Shown in a SARS-CoV-2 protease structure model study to potentially bind and inhibit the papain like viral protease (PLVP) of SARS-CoV-2.
Catalog No. 50-205-4091