Learn More
TARGETMOL CHEMICALS INC DARUNAVIR 50MG
SDP
Catalog No. 502577160
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Darunavir (TMC114) is an HIV PROTEASE INHIBITOR that is used in the treatment of AIDS and HIV INFECTIONS. Due to the emergence of ANTIVIRAL DRUG RESISTANCE when used alone it is administered in combination with other ANTI-HIV AGENTS. purity: 99%
Catalog No. 50-257-7160 Supplier TARGETMOL CHEMICALS INC Supplier No. T232450MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Darunavir (TMC114) is an HIV PROTEASE INHIBITOR that is used in the treatment of AIDS and HIV INFECTIONS. Due to the emergence of ANTIVIRAL DRUG RESISTANCE when used alone it is administered in combination with other ANTI-HIV AGENTS. purity: 99%