Learn More
TARGETMOL CHEMICALS INC DARUNAVIR ETHANOLATE 50MG
SDP
Catalog No. 502580313
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Darunavir Ethanolate (UIC 94017) is an HIV PROTEASE INHIBITOR that is used in the treatment of AIDS and HIV INFECTIONS. Due to the emergence of ANTIVIRAL DRUG RESISTANCE when used alone it is administered in combination with other ANTI-HIV AGENTS. purity: 99%
Catalog No. 50-258-0313 Supplier TARGETMOL CHEMICALS INC Supplier No. T333550MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Darunavir Ethanolate (UIC 94017) is an HIV PROTEASE INHIBITOR that is used in the treatment of AIDS and HIV INFECTIONS. Due to the emergence of ANTIVIRAL DRUG RESISTANCE when used alone it is administered in combination with other ANTI-HIV AGENTS. purity: 99%