Learn More
Medchemexpress LLC HY-17041 5mg Medchemexpress, Darunavir (Ethanolate) CAS:635728-49-3 Purity:>98%
SDP
Catalog No. 502030759
Encompass_Preferred
Medchemexpress, HY-17041 5mg Darunavir (Ethanolate) CAS:635728-49-3 Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0759 Supplier Medchemexpress LLC Supplier No. HY170415MG
Medchemexpress, HY-17041 5mg Darunavir (Ethanolate) CAS:635728-49-3 Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.