Learn More
Selleck Chemical LLC Darunavir Ethanolate S1620-5mg
SDP
Catalog No. 503108492
Encompass_Preferred
Darunavir Ethanolate (TMC-114 UIC 94017) is a nonpeptidic HIV protease inhibitor used to treat HIV infection
Catalog No. 50-310-8492 Supplier Selleck Chemical LLC Supplier No. S16205MG
Darunavir Ethanolate (TMC-114 UIC 94017) is a nonpeptidic HIV protease inhibitor used to treat HIV infection