Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Darunavir, Thermo Scientific™

Catalog No. AC469131000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469131000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC469131000 Supplier Thermo Scientific Chemicals Supplier No. 469131000
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 206361-99-1
Molecular Formula C27H37N3O7S
Molecular Weight (g/mol) 547.67
MDL Number MFCD09260006
InChI Key CJBJHOAVZSMMDJ-HEXNFIEUSA-N
IUPAC Name (3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
SMILES CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CO[C@H]2OCC[C@@H]12)S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White
Quantity 100 mg
Formula Weight 547.67
Chemical Name or Material Darunavir
Hazard Category GHS07: Exclamation mark
ShelfLife 2 years
Recommended Storage Freezer -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.