Learn More
Cayman Chemical Darunavir (ethanolate), Purity: ≥95%, CAS: 635728-49-3, 10 mg
SDP
Catalog No. 502990120
Encompass_Preferred
An HIV-1 protease inhibitor; active against HIV-1LAI in MT-2 cells (IC50 = 3 nM) (CC50 = 74.4 μM); active against wild-type and multidrug-resistant clinical isolates of HIV-1 in PHA-PBMCs (IC50s = 3 and 3-29 nM, respectively); inhibits cell-free diffusion and cell-to-cell spread of HIV-1 in Jurkat cell populations (IC50s = 2.5 and 2.8 nM, respectively)
Catalog No. 50-299-0120 Supplier Cayman Chemical Supplier No. 3108310 MG
An HIV-1 protease inhibitor; active against HIV-1LAI in MT-2 cells (IC50 = 3 nM) (CC50 = 74.4 μM); active against wild-type and multidrug-resistant clinical isolates of HIV-1 in PHA-PBMCs (IC50s = 3 and 3-29 nM, respectively); inhibits cell-free diffusion and cell-to-cell spread of HIV-1 in Jurkat cell populations (IC50s = 2.5 and 2.8 nM, respectively)