Learn More
Medchemexpress LLC HY-15404 5mg Medchemexpress, Darusentan CAS:171714-84-4 Purity:>98%
SDP
Catalog No. 501932734
Encompass_Preferred
Medchemexpress, HY-15404 5mg Darusentan CAS:171714-84-4 Darusentan is a selective endothelin A ( ETA ) receptor antagonist. Darusentan competes for radiolabeled endothelin binding in rat aortic vascular smooth muscle cells (RAVSMs) membranes with single-site kinetics, exhibiting a K i =13 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2734 Supplier Medchemexpress LLC Supplier No. HY154045MG
Medchemexpress, HY-15404 5mg Darusentan CAS:171714-84-4 Darusentan is a selective endothelin A ( ETA ) receptor antagonist. Darusentan competes for radiolabeled endothelin binding in rat aortic vascular smooth muscle cells (RAVSMs) membranes with single-site kinetics, exhibiting a K i =13 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.