Learn More
Medchemexpress LLC Darutoside | 59219-65-7 | 5 MG
SDP
Catalog No. 5000419391
Encompass_Preferred
Darutoside is an orally effective diterpene compound with significant anti-inflammatory, analgesic, wound healing promotion, and immunomodulatory activities. It reduces edema and pain by inhibiting COX-2 expression and inflammatory cell migration, and regulates macrophage polarization towards the M2 type by inhibiting the NF-κB pathway, which alleviates inflammation and promotes wound healing. Additionally, it significantly alleviates acute gouty arthritis through multi-target regulation of metabolic networks.
  • Inhibits COX-2 expression
  • Inhibits NF-κB pathway
  • Regulates macrophage polarization towards the M2 type
  • Reduces edema and pain
  • Promotes wound healing
  • Alleviates acute gouty arthritis
Catalog No. 50-004-19391 Supplier Medchemexpress LLC Supplier No. HYN60285MG
Darutoside is an orally effective diterpene compound with significant anti-inflammatory, analgesic, wound healing promotion, and immunomodulatory activities. It reduces edema and pain by inhibiting COX-2 expression and inflammatory cell migration, and regulates macrophage polarization towards the M2 type by inhibiting the NF-κB pathway, which alleviates inflammation and promotes wound healing. Additionally, it significantly alleviates acute gouty arthritis through multi-target regulation of metabolic networks.
  • Inhibits COX-2 expression
  • Inhibits NF-κB pathway
  • Regulates macrophage polarization towards the M2 type
  • Reduces edema and pain
  • Promotes wound healing
  • Alleviates acute gouty arthritis