Learn More
Medchemexpress LLC Darutoside | 59219-65-7 | MFCD17214820 | 99.9% | 484.62 g/mol | C26H44O8 | 10 MG
SDP
Catalog No. 5000289959
Encompass_Preferred
Darutoside is a diterpenoid glycoside supplied as a high-purity analytical standard for laboratory research. It is provided as a powdered material with documentation for purity and safety, and is intended for research use only (not for human or animal therapeutic use).
  • High purity suitable for analytical and research applications.
  • Molecular formula C26H44O8 and molecular weight 484.62 g/mol.
  • Documented CAS number and certificate of analysis available.
  • Packaged in small analytical quantities (for example, 10 mg) to support assay and reference work.
  • Store sealed at 4°C, protected from moisture and light; solvent storage guidelines apply for longer term.
Catalog No. 50-002-89959 Supplier Medchemexpress LLC Supplier No. HYN602810MG
Darutoside is a diterpenoid glycoside supplied as a high-purity analytical standard for laboratory research. It is provided as a powdered material with documentation for purity and safety, and is intended for research use only (not for human or animal therapeutic use).
  • High purity suitable for analytical and research applications.
  • Molecular formula C26H44O8 and molecular weight 484.62 g/mol.
  • Documented CAS number and certificate of analysis available.
  • Packaged in small analytical quantities (for example, 10 mg) to support assay and reference work.
  • Store sealed at 4°C, protected from moisture and light; solvent storage guidelines apply for longer term.