Learn More
Cayman Chemical DASA-58, 1203494-49-8, 10 mg
SDP
Supplier:  Cayman Chemical 1394110
Encompass_Preferred
A selective activator of PKM2 (AC50 = 38 nM in vitro, EC50 = 19.6 µM in cells); converts PKM2 to a constitutively active enzyme state that is resistant to inhibition by tyrosine-phosphorylated proteins
Catalog No. NC1401444