Learn More
Selleck Chemical LLC Dasabuvir (ABT-333) 10mg 1132935-63-7
SDP
Supplier:  Selleck Chemical LLC S540210MG
Encompass_Preferred
Dasabuvir (ABT-333) is a non-nucleoside inhibitor of NS5B viral RNA-dependent RNA polymerase that inhibits recombinant NS5B polymerases derived from HCV genotype 1a and 1b clinical isolates with IC50 values between 2.2 and 10.7 nM. It is at least 7,000-fold selective for the inhibition of HCV genotype 1 polymerases over human/mammalian polymerases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8411