Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasantafil | 569351-91-3 | 98.4% | 522.39 g/mol | C22H28BrN5O5 | 5 MG
SDP

Catalog No. 5000242896
Encompass_Preferred

Dasantafil (SCH-446132) is a small-molecule phosphodiesterase-5A (PDE5A) inhibitor used in preclinical research of genitourinary disorders, including studies of erectile dysfunction. The compound is supplied as a solid with high chemical purity and includes recommended solubility and storage guidance suitable for in vitro and in vivo formulation.

  • High purity: 98.35%.
  • Selective PDE5A inhibitor for mechanistic and pharmacology studies.
  • Molecular weight 522.39 g/mol.
  • Soluble in DMSO (≥100 mg/mL); in vivo formulations yield clear solutions ≥2.5 mg/mL.
  • Storage recommendations: sealed, away from moisture; in solvent -80°C (6 months), -20°C (1 month).
  • Supplied in small research quantities (for example, 5 mg).

Catalog No. 50-002-42896 Supplier Medchemexpress LLC Supplier No. HY106230A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasantafil (SCH-446132) is a small-molecule phosphodiesterase-5A (PDE5A) inhibitor used in preclinical research of genitourinary disorders, including studies of erectile dysfunction. The compound is supplied as a solid with high chemical purity and includes recommended solubility and storage guidance suitable for in vitro and in vivo formulation.

  • High purity: 98.35%.
  • Selective PDE5A inhibitor for mechanistic and pharmacology studies.
  • Molecular weight 522.39 g/mol.
  • Soluble in DMSO (≥100 mg/mL); in vivo formulations yield clear solutions ≥2.5 mg/mL.
  • Storage recommendations: sealed, away from moisture; in solvent -80°C (6 months), -20°C (1 month).
  • Supplied in small research quantities (for example, 5 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.