Learn More
Eurofins Discoverx Dasatinib, 10 mg. (92-1183)
Supplier:  Eurofins Discoverx 921183
Encompass
Dasatinib (Sprycel, BMS-354825) is an oral BCR-ABL, c-KIT, SRC inhibitor (IC50=<1, 79, and 0.8, respectively) and cancer drug developed by Bristol-Myers Squibb. It is most commonly used for treatment of chronic myelogenous leukemia (CML) and Philadelphia chromosome-positive acute lymphoblastic leukemia (Ph+ ALL). Inhibits tyrosine kinases via competing for ATP. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9711