Learn More
Medchemexpress LLC HY-10181 200mg Medchemexpress, Dasatinib CAS:302962-49-8 Purity:>98%
SDP
Catalog No. 502028682
Encompass_Preferred
Medchemexpress, HY-10181 200mg Dasatinib CAS:302962-49-8 Dasatinib (BMS-354825) is a potent and orally active dual Bcr-Abl and Src family tyrosine kinase inhibitor with IC50s of 0.6 nM, 0.8 nM, respectively. Dasatinib also inhibits Abl, Src, Fyn, c-Kit and c-KitD816V with IC50s of 2.8 nM, 79 nM and 37 nM, respectively. Dasatinib also induces apoptosis and autophagy. Dasatinib shows potently antitumor activity and has the potential for chronic myelogenous leukemia (CML) treatment[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8682 Supplier Medchemexpress LLC Supplier No. HY10181200MG
Medchemexpress, HY-10181 200mg Dasatinib CAS:302962-49-8 Dasatinib (BMS-354825) is a potent and orally active dual Bcr-Abl and Src family tyrosine kinase inhibitor with IC50s of 0.6 nM, 0.8 nM, respectively. Dasatinib also inhibits Abl, Src, Fyn, c-Kit and c-KitD816V with IC50s of 2.8 nM, 79 nM and 37 nM, respectively. Dasatinib also induces apoptosis and autophagy. Dasatinib shows potently antitumor activity and has the potential for chronic myelogenous leukemia (CML) treatment[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.