Learn More
AXON MEDCHEM LLC DASATINIB 2X10MG
SDP
Catalog No. NC1773701
Encompass
Dasatinib - BMS 354825 | Sprycel  Based on 18 reference(s) in Google Scholar Axon 1392CAS [302962-49-8]MF C22H26ClN7O2SMW 488.01 Purity:99% Soluble in DMSO Dasatinib
Catalog No. NC1773701 Supplier AXON MEDCHEM LLC Supplier No. 13922X10MG
Dasatinib - BMS 354825 | Sprycel  Based on 18 reference(s) in Google Scholar Axon 1392CAS [302962-49-8]MF C22H26ClN7O2SMW 488.01 Purity:99% Soluble in DMSO Dasatinib