Learn More
Medchemexpress LLC HY-10181 500mg , Dasatinib BMS-354825 CAS:302962-49-8 Purity:98%
SDP
Catalog No. 501492321
Encompass_Preferred
Medchemexpress, HY-10181 500mg Dasatinib BMS-354825 CAS:302962-49-8 Formula:C22H26ClN7O2S IC50: 0.6 nM/0.8 nM (Abl WT /Src) IC50: 79 nM/37 nM (c-Kit WT /c-Kit D816V ) Purity:98% Dasatinib is a potent and dual Abl WT / Src inhibitor IC 50 of 0.6 nM/0.8 nM respectively; also inhibits c-Kit WT / c-Kit D816V with IC 50 of 79 nM/37 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2321 Supplier Medchemexpress LLC Supplier No. HY10181500MG
Medchemexpress, HY-10181 500mg Dasatinib BMS-354825 CAS:302962-49-8 Formula:C22H26ClN7O2S IC50: 0.6 nM/0.8 nM (Abl WT /Src) IC50: 79 nM/37 nM (c-Kit WT /c-Kit D816V ) Purity:98% Dasatinib is a potent and dual Abl WT / Src inhibitor IC 50 of 0.6 nM/0.8 nM respectively; also inhibits c-Kit WT / c-Kit D816V with IC 50 of 79 nM/37 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.