Learn More
Apexbio Technology LLC Dasatinib (BMS-354825), 302962-49-8, MFCD11046566, 100 mg
SDP
Catalog No. 501013591
Encompass_Preferred
MF: C22H26ClN7O2S, MW: 488.01, Solubility: 24.4mg/mL in DMSO. Dasatinib is a small-molecule inhibitor of both the Src and Bcr-Abl tyrosine kinases with IC50 values of 0.5 nM and 1 nM.
Catalog No. 50-101-3591 Supplier Apexbio Technology LLC Supplier No. A3017100
MF: C22H26ClN7O2S, MW: 488.01, Solubility: 24.4mg/mL in DMSO. Dasatinib is a small-molecule inhibitor of both the Src and Bcr-Abl tyrosine kinases with IC50 values of 0.5 nM and 1 nM.