Learn More
Apexbio Technology LLC Dasatinib (BMS-354825), 10mM (in 1mL DMSO) Cas No: 302962-49-8 MFCD: MFCD11046566
SDP
Catalog No. NC1233578
Encompass_Preferred
Dasatinib is a small-molecule inhibitor of both the Src and Bcr-Abl tyrosine kinases with IC50values of 0.5 nM and 1 nM. The oncogenic tyrosine kinase Bcr-Abl, plays a critical role in the pathogenesis of CML(chronic myelogenous leukemia). There are other sizes available. Please inqury us for quote.
Catalog No. NC1233578 Supplier Apexbio Technology LLC Supplier No. A30175.1
Dasatinib is a small-molecule inhibitor of both the Src and Bcr-Abl tyrosine kinases with IC50values of 0.5 nM and 1 nM. The oncogenic tyrosine kinase Bcr-Abl, plays a critical role in the pathogenesis of CML(chronic myelogenous leukemia). There are other sizes available. Please inqury us for quote.