Learn More
Medchemexpress LLC Dasatinib-d8 | 1132093-70-9 | MFCD15071210 | 99.2% | 496.05 g/mol | C22H18D8ClN7O2S | 1 MG
SDP
Catalog No. 5000246169
Encompass_Preferred
Dasatinib-d8 is a deuterium-labeled analogue of dasatinib supplied as a high-purity reference standard for analytical and research use, primarily as an internal standard for quantitative mass spectrometry. It is intended for method development, validation, and stability studies where isotopic labeling improves accuracy and precision (CAS 1132093-70-9).
  • Deuterium-labeled internal standard for mass spectrometry applications.
  • High purity for reliable quantification and reproducible results.
  • Suitable for LC-MS and GC-MS method development and validation.
  • Soluble in DMSO at high concentration for convenient stock preparation.
  • Packaged for analytical reference and stability testing.
Catalog No. 50-002-46169 Supplier Medchemexpress LLC Supplier No. HY10181S1MG
Dasatinib-d8 is a deuterium-labeled analogue of dasatinib supplied as a high-purity reference standard for analytical and research use, primarily as an internal standard for quantitative mass spectrometry. It is intended for method development, validation, and stability studies where isotopic labeling improves accuracy and precision (CAS 1132093-70-9).
  • Deuterium-labeled internal standard for mass spectrometry applications.
  • High purity for reliable quantification and reproducible results.
  • Suitable for LC-MS and GC-MS method development and validation.
  • Soluble in DMSO at high concentration for convenient stock preparation.
  • Packaged for analytical reference and stability testing.