Learn More
LC Laboratories Dasatinib, 302962-49-8, MFCD11046566, 5 g
Supplier:  LC Laboratories D33075G
Encompass
Molecular Formula: C22 H26 Cl N7 O2 S, Molecular Weight: 488.013, >99%, Synonyms: BMS-354825, Store at or below -20 ºC.
Catalog No. NC0488801