Learn More
Medchemexpress LLC HY-10181A 100mg , Dasatinib (hydrochloride) CAS:854001-07-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10181A100MG
Encompass_Preferred
HY-10181A 100mg Dasatinib (hydrochloride) CAS: 854001-07-3 Dasatinib (BMS-354825) hydrochloride is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity. The Kis are 16 pM and 30 pM for Src and Bcr-Abl, respectively. Dasatinib hydrochloride inhibits Bcr-Abl and Src with IC50s of <1.0 nM and 0.5 nM, respectively. Dasatinib hydrochloride also induces apoptosis and autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0414