Learn More
Selleck Chemical LLC Dasatinib hydrochloride 25mg 854001-07-3 BMS-354825 HCl
SDP
Supplier:  Selleck Chemical LLC S525425MG
Encompass_Preferred
Dasatinib hydrochloride (BMS-354825) is the hydrochloride salt form of dasatinib, an inhibitor that targets Abl, Src and c-Kit, with IC50 of <1 nM, 0.8 nM and 79 nM in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8298