Learn More
Selleck Chemical LLC Dasatinib hydrochloride S5254-10mM/1mL
SDP
Catalog No. 503112415
Encompass_Preferred
Dasatinib hydrochloride (BMS-354825) is the hydrochloride salt form of dasatinib an inhibitor that targets Abl Src and c-Kit with IC50 of 1 nM 0 8 nM and 79 nM in cell-free assays respectively
Catalog No. 50-311-2415 Supplier Selleck Chemical LLC Supplier No. S525410MM/1ML
Dasatinib hydrochloride (BMS-354825) is the hydrochloride salt form of dasatinib an inhibitor that targets Abl Src and c-Kit with IC50 of 1 nM 0 8 nM and 79 nM in cell-free assays respectively