Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasatinib hydrochloride | 854001-07-3 | 99.3% | 524.47 g/mol | C22H27Cl2N7O2S | 500 MG
SDP

Catalog No. 5000248133
Encompass_Preferred

Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor used in cancer research and cell signaling studies. It is supplied as a high-purity research reagent intended for in vitro and in vivo kinase inhibition assays and related biochemical applications.

  • Potent ATP-competitive inhibitor of Src and Bcr-Abl kinases.
  • Suitable for in vitro and in vivo research applications.
  • Hydrochloride salt for improved solubility and handling.
  • High purity (99.31% by HPLC) for reliable experimental results.
  • Molecular formula C22H27Cl2N7O2S; molecular weight 524.47 g/mol.
  • Available in a 500 mg research package.

Catalog No. 50-002-48133 Supplier Medchemexpress LLC Supplier No. HY10181A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor used in cancer research and cell signaling studies. It is supplied as a high-purity research reagent intended for in vitro and in vivo kinase inhibition assays and related biochemical applications.

  • Potent ATP-competitive inhibitor of Src and Bcr-Abl kinases.
  • Suitable for in vitro and in vivo research applications.
  • Hydrochloride salt for improved solubility and handling.
  • High purity (99.31% by HPLC) for reliable experimental results.
  • Molecular formula C22H27Cl2N7O2S; molecular weight 524.47 g/mol.
  • Available in a 500 mg research package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.