Learn More
Medchemexpress LLC Dasatinib hydrochloride | 854001-07-3 | 99.3% | 524.5 g·mol⁻¹ | C22H27Cl2N7O2S | 200 MG
SDP
Catalog No. 5000248863
Encompass_Preferred
Dasatinib hydrochloride is a research-grade, ATP-competitive dual Src and Bcr-Abl kinase inhibitor supplied as the hydrochloride salt. It exhibits subnanomolar to low-nanomolar activity against its targets and is intended for laboratory and preclinical research applications; supplied at high purity for analytical use.
  • High purity: 99.31%.
  • Potent dual Src and Bcr-Abl inhibition at subnanomolar to low-nanomolar concentrations.
  • Supplied as the hydrochloride salt in solid form for research use.
  • Suitable for research and analytical applications; not for human consumption.
  • Available in multiple pack sizes, including 200 MG.
Catalog No. 50-002-48863 Supplier Medchemexpress LLC Supplier No. HY10181A200MG
Dasatinib hydrochloride is a research-grade, ATP-competitive dual Src and Bcr-Abl kinase inhibitor supplied as the hydrochloride salt. It exhibits subnanomolar to low-nanomolar activity against its targets and is intended for laboratory and preclinical research applications; supplied at high purity for analytical use.
  • High purity: 99.31%.
  • Potent dual Src and Bcr-Abl inhibition at subnanomolar to low-nanomolar concentrations.
  • Supplied as the hydrochloride salt in solid form for research use.
  • Suitable for research and analytical applications; not for human consumption.
  • Available in multiple pack sizes, including 200 MG.