Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasatinib hydrochloride | 854001-07-3 | MFCD00713745 | 99.3% | 524.5 g/mol | C22H27Cl2N7O2S | 50 MG
SDP

Catalog No. 5000248864
Encompass_Preferred

Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor supplied as a purified solid for research and analytical applications. It is intended for in vitro studies and signal transduction research; not for human or clinical use.

  • High purity (99.3%).
  • Suitable for research and analytical applications.
  • Molecular formula C22H27Cl2N7O2S; molecular weight 524.5 g/mol.
  • Registered CAS number 854001-07-3 for unambiguous identification.
  • Available in small milligram pack sizes for laboratory use.

Catalog No. 50-002-48864 Supplier Medchemexpress LLC Supplier No. HY10181A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor supplied as a purified solid for research and analytical applications. It is intended for in vitro studies and signal transduction research; not for human or clinical use.

  • High purity (99.3%).
  • Suitable for research and analytical applications.
  • Molecular formula C22H27Cl2N7O2S; molecular weight 524.5 g/mol.
  • Registered CAS number 854001-07-3 for unambiguous identification.
  • Available in small milligram pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.