Learn More
Medchemexpress LLC Dasatinib hydrochloride | 854001-07-3 | MFCD26960525 | 99.3% | 524.5 g/mol | C22H27Cl2N7O2S | 1 ML
SDP
Catalog No. 5000248116
Encompass_Preferred
Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive inhibitor of Src and Bcr-Abl family tyrosine kinases used in preclinical research. It is typically supplied as a pre-dissolved 10 mM solution in DMSO for in vitro assays (CAS 854001-07-3).
  • Potent ATP-competitive inhibitor of Src and Bcr-Abl kinases.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity suitable for analytical and biological research (approx. 99.3%).
  • Useful for in vitro kinase inhibition and cell signaling studies.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-48116 Supplier Medchemexpress LLC Supplier No. HY10181A10MM 1ML
Dasatinib hydrochloride is the hydrochloride salt of dasatinib, a potent ATP-competitive inhibitor of Src and Bcr-Abl family tyrosine kinases used in preclinical research. It is typically supplied as a pre-dissolved 10 mM solution in DMSO for in vitro assays (CAS 854001-07-3).
  • Potent ATP-competitive inhibitor of Src and Bcr-Abl kinases.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • High purity suitable for analytical and biological research (approx. 99.3%).
  • Useful for in vitro kinase inhibition and cell signaling studies.
  • For research use only; not for human or veterinary use.