Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasatinib intermediate-1 | 302964-08-5 | ≥98.0% | 394.28 | 100 G
SDP

Supplier:  Medchemexpress LLC HY41940100G

Encompass_Preferred

Dasatinib intermediate-1 is an intermediate in the synthesis of Dasatinib, a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity.

  • Intermediate in Dasatinib synthesis.
  • Targets Bcr-Abl.
  • Intended for research use only.
  • Store at -20°C, sealed, away from moisture.
  • When in solvent, stable for 6 months at -80°C or 1 month at -20°C.

Catalog No. 50-004-03545


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.