Learn More
Medchemexpress LLC HY-131669 5mg Medchemexpress, Dasatinib metabolite M6 CAS:910297-53-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1316695MG
Encompass_Preferred
Medchemexpress, HY-131669 5mg Dasatinib metabolite M6 CAS:910297-53-9 Dasatinib metabolite M6 (Dasatinib carboxylic acid) is an oxidative metabolite of Dasatinib (HY-10181). Dasatinib is a potent and orally active dual Bcr-Abl and Src family tyrosine kinase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1294