Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasatinib monohydrate | 863127-77-9 | MFCD08704581 | 99.6% | 506.0 g/mol | C22H28ClN7O3S | 100 MG
SDP

Catalog No. 5000248117
Encompass_Preferred

Dasatinib monohydrate is the monohydrate form of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor used in preclinical and biochemical research. Supplied as a purified solid, it is commonly used in cell signaling and oncology-related assays where selective kinase inhibition is required.

  • High purity (99.6%).
  • Molecular weight 506.0 g/mol.
  • Chemical formula C22H28ClN7O3S.
  • Supplied as a 100 MG solid for laboratory use.
  • Suitable for preclinical research and biochemical assays.
  • Available in multiple pack sizes to support experimental scaling.

Catalog No. 50-002-48117 Supplier Medchemexpress LLC Supplier No. HY10181B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasatinib monohydrate is the monohydrate form of dasatinib, a potent ATP-competitive dual Src/Bcr-Abl kinase inhibitor used in preclinical and biochemical research. Supplied as a purified solid, it is commonly used in cell signaling and oncology-related assays where selective kinase inhibition is required.

  • High purity (99.6%).
  • Molecular weight 506.0 g/mol.
  • Chemical formula C22H28ClN7O3S.
  • Supplied as a 100 MG solid for laboratory use.
  • Suitable for preclinical research and biochemical assays.
  • Available in multiple pack sizes to support experimental scaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.