Learn More
Medchemexpress LLC Dasatinib monohydrate | 863127-77-9 | MFCD08704581 | 99.6% | 506.02 g/mol | C22H28ClN7O3S | 1 ML
SDP
Catalog No. 5000246821
Encompass_Preferred
Dasatinib monohydrate is a potent, ATP-competitive dual Src and Bcr-Abl tyrosine kinase inhibitor supplied for in vitro oncology research. Available in solution and solid forms, it is intended for biochemical and cell-based assay applications where a well-characterized kinase inhibitor is required.
  • High purity suitable for research applications (manufacturer lists ~99.6% purity).
  • Provided as a 10 mM solution in DMSO for ready-to-use dosing.
  • Also available as solid material for custom formulation or storage.
  • Potent Src and Bcr-Abl inhibition supports signaling and mechanistic studies.
  • Compatible with biochemical and cell-based assay formats.
  • Documented analytical characterization for reproducible results.
Catalog No. 50-002-46821 Supplier Medchemexpress LLC Supplier No. HY10181B10MM 1ML
Dasatinib monohydrate is a potent, ATP-competitive dual Src and Bcr-Abl tyrosine kinase inhibitor supplied for in vitro oncology research. Available in solution and solid forms, it is intended for biochemical and cell-based assay applications where a well-characterized kinase inhibitor is required.
  • High purity suitable for research applications (manufacturer lists ~99.6% purity).
  • Provided as a 10 mM solution in DMSO for ready-to-use dosing.
  • Also available as solid material for custom formulation or storage.
  • Potent Src and Bcr-Abl inhibition supports signaling and mechanistic studies.
  • Compatible with biochemical and cell-based assay formats.
  • Documented analytical characterization for reproducible results.