Learn More
Medchemexpress LLC Dasatinib monohydrate | 863127-77-9 | MFCD08704581 | 99.6% | 506.03 g/mol | C22H26ClN7O2S · H2O | 50 MG
SDP
Catalog No. 5000248097
Encompass_Preferred
Dasatinib monohydrate is a potent, ATP-competitive dual Src and Bcr-Abl tyrosine kinase inhibitor used in preclinical oncology research. Provided in a defined monohydrate form, it is supplied as a solid and as a DMSO solution for in vitro assay use, enabling reproducible pharmacology and biochemical studies.
  • High purity: 99.6% (reported), suitable for research applications.
  • Monohydrate form for defined solid-state properties and reproducible handling.
  • Potent dual Src and Bcr-Abl inhibition for kinase signaling and cancer biology studies.
  • Available as 50 MG solid and as a 10 mM solution in DMSO for assay flexibility.
  • Molecular weight approximately 506.03 g/mol and CAS 863127-77-9 for unambiguous identification.
Catalog No. 50-002-48097 Supplier Medchemexpress LLC Supplier No. HY10181B50MG
Dasatinib monohydrate is a potent, ATP-competitive dual Src and Bcr-Abl tyrosine kinase inhibitor used in preclinical oncology research. Provided in a defined monohydrate form, it is supplied as a solid and as a DMSO solution for in vitro assay use, enabling reproducible pharmacology and biochemical studies.
  • High purity: 99.6% (reported), suitable for research applications.
  • Monohydrate form for defined solid-state properties and reproducible handling.
  • Potent dual Src and Bcr-Abl inhibition for kinase signaling and cancer biology studies.
  • Available as 50 MG solid and as a 10 mM solution in DMSO for assay flexibility.
  • Molecular weight approximately 506.03 g/mol and CAS 863127-77-9 for unambiguous identification.