Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasatinib N-oxide | 910297-52-8 | MFCD15071211 | 99.2% | 504.00 g/mol | C22H26ClN7O3S | 5 MG
SDP

Catalog No. 5000262580
Encompass_Preferred

Dasatinib N-oxide is an analytical standard and a minor oxidative metabolite of dasatinib, a dual Src/Bcr-Abl tyrosine kinase inhibitor. Supplied as a solid reference material, it is intended for research and analytical applications such as metabolite identification, method development, and quantitative assays. Not for human or veterinary use.

  • Acts as a reference standard for analytical and metabolite studies.
  • High reported purity suitable for analytical work.
  • Molecular weight 504.00 g/mol; formula C22H26ClN7O3S.
  • Supplied as a solid for convenient storage and handling.
  • Storage: protect from light; 4°C recommended; in solution store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only, not for clinical or diagnostic use.

Catalog No. 50-002-62580 Supplier Medchemexpress LLC Supplier No. HY1337945MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasatinib N-oxide is an analytical standard and a minor oxidative metabolite of dasatinib, a dual Src/Bcr-Abl tyrosine kinase inhibitor. Supplied as a solid reference material, it is intended for research and analytical applications such as metabolite identification, method development, and quantitative assays. Not for human or veterinary use.

  • Acts as a reference standard for analytical and metabolite studies.
  • High reported purity suitable for analytical work.
  • Molecular weight 504.00 g/mol; formula C22H26ClN7O3S.
  • Supplied as a solid for convenient storage and handling.
  • Storage: protect from light; 4°C recommended; in solution store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only, not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.