Learn More
Medchemexpress LLC Dasatinib N-oxide | 910297-52-8 | MFCD15071211 | 99.2% | C22H26ClN7O3S | 10MG
SDP
Catalog No. 5000259917
Encompass_Preferred
Dasatinib N-oxide is a minor oxidative metabolite of the kinase inhibitor dasatinib, provided as a high-purity small-molecule reference standard for research. It is intended for use in metabolic profiling, pharmacokinetic studies, impurity identification, and analytical method development.
  • High purity suitable for use as an analytical reference standard.
  • Verified identity with molecular formula C22H26ClN7O3S.
  • Applicable to metabolism and pharmacokinetic studies.
  • Compatible with LC-MS, HPLC, and related analytical techniques.
  • Supplied in milligram-scale quantities for laboratory research.
Catalog No. 50-002-59917 Supplier Medchemexpress LLC Supplier No. HY13379410MG
Dasatinib N-oxide is a minor oxidative metabolite of the kinase inhibitor dasatinib, provided as a high-purity small-molecule reference standard for research. It is intended for use in metabolic profiling, pharmacokinetic studies, impurity identification, and analytical method development.
  • High purity suitable for use as an analytical reference standard.
  • Verified identity with molecular formula C22H26ClN7O3S.
  • Applicable to metabolism and pharmacokinetic studies.
  • Compatible with LC-MS, HPLC, and related analytical techniques.
  • Supplied in milligram-scale quantities for laboratory research.