Learn More
MSC Value Collection ZMSCH70003 1/16" ID x 3/16" OD Dash 003 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. NC1898854
Encompass
Value Collection ZMSCH70003 1/16" ID x 3/16" OD Dash 003 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F
Catalog No. NC1898854 Supplier MSC Supplier No. 09260035
Value Collection ZMSCH70003 1/16" ID x 3/16" OD Dash 003 Buna-N O-Ring 1/16" Thick, Round Cross Section, Shore 65 - 75, -30 to 250°F