Learn More
Medchemexpress LLC Dasiglucagon acetate | 1544300-84-6 | 98.7% | 3441.67 | 5 MG
SDP
Catalog No. 5000396279
Encompass_Preferred
Dasiglucagon acetate is a human glucagon analog that is used to increase plasma glucose levels. It is suitable for research related to hypoglycemia.
  • Human glucagon analog
  • Increases plasma glucose
  • Used in hypoglycemia research
Catalog No. 50-003-96279 Supplier Medchemexpress LLC Supplier No. HYP3469A5MG
Dasiglucagon acetate is a human glucagon analog that is used to increase plasma glucose levels. It is suitable for research related to hypoglycemia.
  • Human glucagon analog
  • Increases plasma glucose
  • Used in hypoglycemia research