Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dasminapant | 1570231-89-8 | 99.7% | 1157.40 | C60H72N10O10S2 | 10MG
SDP

Catalog No. 5000235772
Encompass_Preferred

Dasminapant (APG-1387) is a bivalent SMAC mimetic that antagonizes inhibitor of apoptosis proteins (IAPs). It induces degradation of cIAP-1 and XIAP, activates caspase-3, and promotes PARP cleavage, making it suitable for in vitro research on apoptosis and cancer biology.

  • High purity of 99.7%.
  • Molecular formula C60H72N10O10S2.
  • Molecular weight 1157.40.
  • Available in small milligram pack sizes suitable for research use.
  • Mechanism: induces cIAP-1 and XIAP degradation and caspase-3 activation.

Catalog No. 50-002-35772 Supplier Medchemexpress LLC Supplier No. HY12559310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dasminapant (APG-1387) is a bivalent SMAC mimetic that antagonizes inhibitor of apoptosis proteins (IAPs). It induces degradation of cIAP-1 and XIAP, activates caspase-3, and promotes PARP cleavage, making it suitable for in vitro research on apoptosis and cancer biology.

  • High purity of 99.7%.
  • Molecular formula C60H72N10O10S2.
  • Molecular weight 1157.40.
  • Available in small milligram pack sizes suitable for research use.
  • Mechanism: induces cIAP-1 and XIAP degradation and caspase-3 activation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.