Learn More
Biotium DASPEI
Catalog No. 501964751
Encompass_Preferred
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) is used to stain mitochondria in live cells.Properties: Ex/ Em = 461/589 nm (MeOH); Store at 4°C and protect from light, especially in solution; C17H21IN2; MW: 380; [3785-01-1]
Catalog No. 50-196-4751 Supplier Biotium Supplier No. 70018
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) is used to stain mitochondria in live cells.Properties: Ex/ Em = 461/589 nm (MeOH); Store at 4°C and protect from light, especially in solution; C17H21IN2; MW: 380; [3785-01-1]