Learn More
Enterprise Technology Solutions Data Accessories Company Monitor Riser Block, Stackable, 12"x12"x1-1/4", Black
SDP
Catalog No. 5000791509
Encompass_Preferred
Use the MP-106 Ergonomic Monitor Riser Block to reduce muscle fatigue and promote better seating posture. Heavy-duty, stackable riser raises the computer monitor in 1-1/4" (3.2cm) increments to an ergonomically comfortable height. Add multiple blocks (not included) to suit your needs. Each TAA-compliant, high-impact plastic block supports monitors weighing up to 77 lb. (35 kg). Block platform measures 12" x 12".
Catalog No. 50-007-91509 Supplier Enterprise Technology Solutions Supplier No. DTA02151
Use the MP-106 Ergonomic Monitor Riser Block to reduce muscle fatigue and promote better seating posture. Heavy-duty, stackable riser raises the computer monitor in 1-1/4" (3.2cm) increments to an ergonomically comfortable height. Add multiple blocks (not included) to suit your needs. Each TAA-compliant, high-impact plastic block supports monitors weighing up to 77 lb. (35 kg). Block platform measures 12" x 12".