Learn More
GENIE GROUP UVA/UVC Light Meter/Datalogger
Catalog No. NC1714249
Encompass_Preferred
The SDL470 is a UVA/UVC Light Meter and Data logger. The SDL470 can measure both long wave and short wave UV irradiance under UVA and UVC light sources respectively. This meter features a cosine correction filter, data logging to Excel format, offset adjustment for relative measurements, type K/J thermocouple input for temperature measurements (probe sold separately), record/recall modes, min and max readings, data hold plus auto power off with disable function. This product comes with a built-in tilt stand, SD memory card, UVA light sensor with protective cover, UVC light sensor, hard case, universal AC adaptor, and 6 x AA batteries.
Catalog No. NC1714249 Supplier GENIE GROUP Supplier No. SDL470NISTL
The SDL470 is a UVA/UVC Light Meter and Data logger. The SDL470 can measure both long wave and short wave UV irradiance under UVA and UVC light sources respectively. This meter features a cosine correction filter, data logging to Excel format, offset adjustment for relative measurements, type K/J thermocouple input for temperature measurements (probe sold separately), record/recall modes, min and max readings, data hold plus auto power off with disable function. This product comes with a built-in tilt stand, SD memory card, UVA light sensor with protective cover, UVC light sensor, hard case, universal AC adaptor, and 6 x AA batteries.