Learn More
Cole-Parmer TSI 9565-X VelociCalc Ventilation Meter Only
Supplier:  Cole-Parmer 1030248
Encompass
These are TSI's most advanced thermoanemometers with Bluetooth printer capability and ability to log up to 26,500 samples, and 100 test IDs. They offer manual or continuous data logging, and display up to five measurements at the same time. They come with NIST-traceable calibration report that are supplied by the manufacturer.
Catalog No. 50-633-413